Anti-Interferon alpha 11 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7023R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Interferon alpha 11
- Antigen symbol:IFNA11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:na11|N alpha 11|interferon alpha-1.|N-alpha-11|Interferon alpha-11|Limitin|N-alpha 11|NAB_MOUSE
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Produced by macrophages, IFN alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Interferon alpha 11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat