Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-ELL2 Rabbit Polyclonal Antibody
Anti-ELL2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6993R
Supplier:  Bioss
undefined
Anti-ELL2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6993R
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    elongation factor, RNA polymerase II, 2
  • Symbol antigenu:
    ELL2
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    liquid
  • Identifikační číslo genu:
    22936
  • Synonyma antigenu:
    Ell2|ELL related RNA polymerase II elongation factor|RNA polymerase II elongation factor ELL2.|Elongation factor RNA polymerase II 2|ELL2_HUMAN
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Koncentrace:
    1 ug/ul
  • Transportní teplota:
    4 °C
  • Čištění:
    Purified by Protein A.
  • Rozměry protilátky:
    100 μg
  • Bal.:
    100 µl

Specifications

About this item

Elongation factor that can increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ELL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat