Anti-GPR171 Rabbit Polyclonal Antibody
BOSSBS-6564R
: Bioss
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 171
- Antigen symbol:GPR171
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:29909
- Antigen synonyms:GP171_HUMAN.|H963|GPR171|G protein coupled receptor 171|GPR 171|Platelet activating receptor homolog|G protein coupled receptor H963
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Platelet Activating Receptor H963 is an orphan receptor that is a member of the Platelet Activating Factor Receptor subfamily. ESTs have been isolated from normal human blood (activated), liver, and testis libraries, and from a human uterine adenocarcinoma library.
WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: GPR171
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat