Anti-Gene 21 protein/NPRL2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6079R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Nitrogen permease regulator like 2
- Antigen symbol:NPRL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:NPR like 2|Tumor suppressor candidate 4|TUSC 4|TUSC4 protein|NPR2 like|Gene 21 protein|NPRL 2|NPR 2L|TUSC4|NPR L2|Homologous to yeast nitrogen permease candidate tumor suppressor|NPRL2_HUMAN.|Homologous to yeast nitrogen permease|G21 protein|TUSC 4 protein|NPRL2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
NPR2L is homologous to yeast nitrogen permease and is a candidate tumor suppressor, being a negative regulator of cell cycle. Most abundant in skeletal muscle, followed by brain, liver, and pancreas, with lower amounts in lung, kidney, placenta, and heart. Expressed in most lung cancer cell lines tested. There are two isoforms, produced by alternative splicing.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Gene 21 protein/NPRL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat