Anti-CSNK1G2 Rabbit Polyclonal Antibody
BOSSBS-5053R
: Bioss
- Antibody type:Primary
- Antigen name:Casein kinase 1, gamma 2
- Antigen symbol:CSNK1G2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:1455
- Antigen synonyms:Casein kinase I isoform gamma 2|CK1g2|KC1G2_HUMAN|Casein kinase 1|CSNK1G 2|CKI gamma 2|gamma 2|CSNK1G2.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Casein kinase 1 gamma 2 (CSNK1G2) is a serine/threonine protein kinase which shows preferential utilization of acidic proteins such as caseins as a substrate. It can phosphorylate a large number of proteins.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CSNK1G2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: