Anti-Parafibromin Rabbit Polyclonal Antibody
Catalog # BOSSBS-1597R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cell division cycle 73
- Antigen symbol:CDC73
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:79577
- Antigen synonyms:FIHP|C1orf28|HRPT2|HYX|HPTJT|HRPT1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
The HRPT2 gene product, parafibromin, is a tumor suppressor protein that is part of the human Paf1 complex. The yeast counterpart to this complex is part of the RNA polymerase II complex as well, and is important for histone modification and connections to posttranscriptional events. Human parafibromin also associates with the RNA polymerase II large subunit.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Parafibromin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat