ContactUsLinkComponent
VWR®, Flat Caps or Plug Caps for VWR® Centrifuge Tubes
VWR®, Flat Caps or Plug Caps for VWR® Centrifuge Tubes
Catalog # 525-0923
Supplier:  VWR Collection
   Low Manufacturing Impact
   Sustainable Packaging
undefined
VWR®, Flat Caps or Plug Caps for VWR® Centrifuge Tubes
Catalog # 525-0923
Supplier:  VWR Collection
   Low Manufacturing Impact
   Sustainable Packaging
Specifications
  • For:
    50 ml centrifuge tubes
  • Description:
    Flat caps
  • Environmentally Preferable:
    Low Manufacturing Impact,
    Sustainable Packaging
  • Pk:
    500
Specifications
About this item
Purple PP. Sustainable design based on virgin resins.
Closures for use with VWR® 50 ml tubes. Non sterile. Not autoclavable.