

Piperine for synthesis, Sigma-Aldrich®
Catalog # 8.21036.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:0.04 g/l (18 °C)
- Melting Point:127 - 130 °C
- Molar Mass:285.34 g/mol
- Assay (HPLC, area%):≥98 % (a/a)
- Identity (IR):Passes test
- Storage temperature:+15...+25 °C
- Synonyms:1-Piperoylpiperidine
Specification Test Results
About this item
:
:285.34 g/mol
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:202-348-0
UN:3077
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:94-62-2
Formula:C₁₇H₁₉NO₃
MDL Number:MFCD00005839