Order Entry
Czech Republic
ContactUsLinkComponent
 
1,4-Dihydro-4-pyridinone for synthesis, Sigma-Aldrich®
Catalog # 8.22140.0050
Supplier:  Merck
CAS Number:  
1,4-Dihydro-4-pyridinone for synthesis, Sigma-Aldrich®
Catalog # 8.22140.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    140 - 147 °C
  • Molar Mass:
    95.1 g/mol
  • Bulk Density:
    600 kg/m3
  • Boiling Point:
    170 - 172 °C (0.03 hPa)
  • Flash Point:
    212 °C
  • pH:
    7.0 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    515 °C
  • Assay (HClO4):
    ≥95%
  • Sulfated ash:
    ≤5%
  • Identity (IR):
    Passes test
  • Storage temperature:
    +15...+25 °C

Specification Test Results

About this item

 :
 :95.1 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-633-2
CAS Number:108-96-3
Formula:C₅H₅NO
MDL Number:MFCD00040458