Order Entry
Czech Republic
ContactUsLinkComponent
 
Health
Irritant
Corrosive
1,3-Dimethyl-2-imidazolidinone ≥99.5%, PESTINORM® for headspace gas chromatography
Catalog # 85448.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Corrosive
1,3-Dimethyl-2-imidazolidinone ≥99.5%, PESTINORM® for headspace gas chromatography
Catalog # 85448.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Identity (IR):
    Complying
  • Purity (GC on anhydrous basis):
    min. 99.5%
  • Water (KF):
    max. 0.03% w/w
  • Color (APHA Pt-Co):
    max. 10
  • GC-HS (Class 1 solvents):
    max. 1 µg/g
  • GC-HS (Class 2 solvents):
    max. 10 µg/g
  • GC-HS (Class 3 solvents):
    max. 50 µg/g
  • GC-HS (DMF):
    max. 20 µg/g
  • GC-HS (Each unknown, as MIBK):
    max. 10 µg/g

Specification Test Results

About this item

CAS Number:80-73-9
 :
Formula:C₅H₁₀N₂O
 :114.15 g/mol
Merck Index:13,03275
Boiling Pt:225.5 °C (1013 hPa)
Melting Pt:8 °C
Density:1.04 g/cm³ (20 °C)
Flash Pt:93 °C
EINECS:201-304-8
UN:2810
ADR:6.1,III
 :GHS08
 :GHS07
 :GHS05
MDL Number:MFCD00003188