ContactUsLinkComponent
Avantor® ACE®, Capillary and Nano Column Coupler
Avantor® ACE®, Capillary and Nano Column Coupler
Catalog # C0003
Supplier:  Avantor
undefined
Avantor® ACE®, Capillary and Nano Column Coupler
Catalog # C0003
Supplier:  Avantor
Specifications
  • Description:
    Capillary column coupler
  • Pk:
    1
Specifications
About this item
Avantor® ACE® capillary/nano column couplers are suitable for connecting capillary and nano LC columns to stand-alone capillary/nano guard cartridges or for making similar LC connections.