Anti-LIMK2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9717
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:LIM domain kinase 2
- Antigen symbol:LIMK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3985
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for LIM domain kinase 2(LIMK2) detection. Tested with WB in Human;Mouse.
Type: Primary
Antigen: LIMK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse