Anti-F3 Rabbit Polyclonal Antibody
Catalog # BSBTPB9701
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:coagulation factor III (thromboplastin, tissue factor)
- Antigen symbol:F3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2152
- Antigen synonyms:TFA|CD142|TF
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Tissue factor(F3) detection. Tested with WB in Human.
Type: Primary
Antigen: F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human