Anti-ADRBK2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9682
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:adrenergic, beta, receptor kinase 2
- Antigen symbol:ADRBK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:157
- Antigen synonyms:BARK2|GRK3
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Beta-adrenergic receptor kinase 2(ADRBK2) detection. Tested with WB in Human.
Type: Primary
Antigen: ADRBK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human