Rat IgG2a kappa Isotype Control [clone: 2A3]
Catalog # 70-4321-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Cross adsorption:No
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Purified Rat IgG2a Isotype Control (2A3)
Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: