Anti-SLC3A2 Rabbit Polyclonal Antibody
ABNOPAB29491
: Abnova
- Antibody type:Primary
- Antigen name:solute carrier family 3 (amino acid transporter heavy chain), member 2
- Antigen symbol:Solute carrier family 3, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:6520
- Antigen synonyms:4F2HC|4F2|NACAE|CD98HC|MDU1|CD98|4T2HC
- Sequence:KGRLDYLSSLKVKGLVLGPIHKNQKDDVAQTDLLQIDPNFGSKEDFDSLLQSAKKKSIRVILDLTPNYRGENSWFSTQVDTVATKVKDALEFWLQAGVDGFQVRDIENLKDASSFLAEWQNITKGFSEDRLLIAGTNSSDL
- Immunogen:Recombinant protein corresponding to amino acids of human SLC3A2.
- Size:100 μl
- Pk:100 µl
SLC3A2 polyclonal antibody raised against recombinant human SLC3A2.
Type: Primary
Antigen: SLC3A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human