Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CDC20 Rabbit Polyclonal Antibody
Anti-CDC20 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29462
Supplier:  Abnova
Anti-CDC20 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29462
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cell division cycle 20
  • Antigen symbol:
    CDC20
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Gene ID:
    991
  • Antigen synonyms:
    CDC20A|p55CDC|bA276H19.3
  • Sequence:
    TPTKKEHQKAWALNLNGFDVEEAKILRLSGKPQNAPEGYQNRLKVLYSQKATPGSSRKTCRYIPSLPDRILDAPEIRNDY
  • Immunogen:
    Recombinant protein corresponding to human CDC20.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against recombinant human CDC20.

Type: Primary
Antigen: CDC20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human