Multi-parameter tester, ColorQ PRO 7
Catalog # LAMO2056
Supplier: LaMotte Chemicals
CAS Number:
Specifications
- Pk:1 KIT
- Description:ColorQ PRO 7
Specifications
About this item
ColorQ PRO 7 is a multi-parameter tester used in pool and spa for water testing. As a hand-held photometer it reads seven test factors (free chlorine, total chlorine, bromine, pH, alkalinity, calcium hardness, and cyanuric acid) directly on a digital display. These seven critical parameters helps to ensure the good quality of water.
- Multiple test parameters
- Greater accuracy than visual comparison testing
- Eliminates the need to visually determine slight colour variations or use look-up tables
Measuring range will be as follows:
Free chlorine (DPD): 0 to 10 ppm
Total chlorine (DPD): 0 to 10 ppm
Bromine (DPD) - uses the chlorine (DPD) reagent set: 0 to 22 ppm
pH: 6,5 to 8,5
Total alkalinity: 0 to 250 ppm
Calcium hardness: 0 to 700 ppm
Cyanuric acid: 0 to 125 ppm
Delivery: ColorQ PRO 7-PLUS meter is supplied with Pool Manager Handbook, saturation index calculator, testing reagents and trilingual instructions (English, Spanish, French) and is packaged in a rugged carrying case for on-site testing. Refill packs of reagents are available.