Orders LinkContactUsLinkComponent
Distillation flasks, engler type
 :   Kavalier
Distillation flasks, engler type
 :  Kavalier
Distillation flasks, engler type
Distillation flasks, engler type
Borosilicate glass 3.3, without reference mark.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.
 : Complies to ASTM E1405 (125 ml) and ASTM D86 (100 ml)
Product Family Options
  • Capacity
    Height
    Ext.Ø
    Neck I-Ø
  • 100 ml
    215 mm
    64 mm
    20 mm
    1 * 1 KS
    1 * 1 KS - 561,00 Kč
    561,00 Kč
    1 * 1 KS
       561,00 Kč
  • 125 ml
    215 mm
    68 mm
    20 mm
    1 * 1 KS
    1 * 1 KS - 731,00 Kč
    731,00 Kč
    1 * 1 KS
       731,00 Kč
  • 250 ml
    215 mm
    85 mm
    20 mm
    1 * 1 KS
    1 * 1 KS