ContactUsLinkComponent
Beakers, Philips, borosilicate glass
Beakers, Philips, borosilicate glass
Katalogové číslo KAVA632411006940
Dodavatel:  Kavalier
Supplier #: 632411006940
undefined
Beakers, Philips, borosilicate glass
Katalogové číslo KAVA632411006940
Dodavatel:  Kavalier
Supplier #: 632411006940
Přihlásit se pro dostupnost
Select an Option
112 mm
Select an Option
1000 ml
Select an Option
184 mm
Select an Option
05
Select an Option
1
O tomto produktu
Borosilicate glass, grade 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Autoclavable
Ideal for general laboratory use. With spout.
Frequently Bought Together