Erlenmeyer flasks with screw cap
: GLASSCO
Borosilicate 3.3 glass, clear, with teflon lined blue PP screw cap.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. Cap temperature resistant up to 140 °C.
Frequently Bought Together