Anti-CD3e/CD4/CD8a Monoclonal Antibody (PE-Cy7®, PE, FITC) [clone: 145-2C11, RM4-5, 53-6.7]
Katalogové číslo 91-0000-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Primary
- Antigen symbol:CD3e, CD4, CD8a
- Clonality:Monoclonal
- Clone:145-2C11, RM4-5, 53-6.7
- Conjugation:PE (Phycoerythrin)-Cy7®/PE (Phycoerythrin)/FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Multiple
- Reactivity:Mouse
- Cross adsorption:No
- Storage temperature:Store undiluted at 2-8 °C, protect from light, do not freeze.
- Size:100 ug
- Pk:100 µG
Specifications
About this item
The Mouse T Lymphocyte Subset backbone panel consists of 3 anti-mouse antibodies, each conjugated to a different fluorochrome. This panel can serve as a backbone for identifying major T cell subsets, and leaves room for additional drop-in markers, if desired. An APC version of this panel is available, where the anti-CD8a antibody is conjugated to APC. Hosts: Armenian Hamster, Rat
Type: Primary
Antigen: CD3e, CD4, CD8a
Clonality: Monoclonal
Clone: 145-2C11, RM4-5, 53-6.7
Conjugation: PE-Cy7®, PE, FITC
Epitope:
Host: Armenian Hamster, Rat
Isotype:
Reactivity: Mouse