Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-HLA-DR Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: L243]
 
Anti-HLA-DR Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: L243]

 

  • Catalog No:
  • 35-9952-T025
  • 35-9952-T500
  • 35-9952-T100
  • Antigen symbol:
  • HLA-DRA
  • HLA-DRA
  • HLA-DRA
  • Antigen name:
  • Major histocompatibility Complex, class II, DR alpha
  • Major histocompatibility Complex, class II, DR alpha
  • Major histocompatibility Complex, class II, DR alpha
  • Antigen synonyms:
  • MLRW|HLA-DRA1
  • MLRW|HLA-DRA1
  • MLRW|HLA-DRA1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • L243
  • L243
  • L243
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 3122
  • 3122
  • 3122
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

 

Frequently Bought Together