Order Entry
Czech Republic
ContactUsLinkComponent
Qualitative filter papers, grades MN 619 and 619 1/4, circles and folded filters
Qualitative filter papers, grades MN 619 and 619 1/4, circles and folded filters
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Qualitative filter papers, grades MN 619 and 619 1/4, circles and folded filters
Supplier:  MACHEREY-NAGEL

Specifications

  • Catalog No:
  • MANA437005
  • MANA437032
  • MANA437011
  • 516-6000
  • 516-6016
  • MANA437012
  • MANA437009
  • MANA537005
  • MANA537018
  • MANA437018
  • MANA437015
  • MANA537011
  • MANA537012
  • MANA437007
  • MANA537007
  • MANA537009
  • MANA537024
  • MANA437024
  • Weight:
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • Thickness:
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • Filtration speed:
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • 100 s (DIN 53137)
  • Ash content:
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%

Specifications

Related Information

Frequently Bought Together