Order Entry
Czech Republic
ContactUsLinkComponent
Qualitative filter paper, grades MN 617 we and 617 we 1/4, circles and folded filters
Qualitative filter paper, grades MN 617 we and 617 we 1/4, circles and folded filters
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Qualitative filter paper, grades MN 617 we and 617 we 1/4, circles and folded filters
Supplier:  MACHEREY-NAGEL

Specifications

  • Catalog No:
  • MANA435007
  • MANA435009
  • MANA435005
  • MANA435018
  • MANA435015
  • MANA435024
  • MANA535005
  • MANA435011
  • MANA535032
  • MANA535018
  • 512-0512
  • MANA535011
  • MANA535012
  • MANA535007
  • MANA435032
  • MANA535009
  • MANA535015
  • MANA535024
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • Particle retention:
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • Filtration speed:
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • 5 s (DIN 53137)
  • Ash content:
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%

Specifications

Related Information

Frequently Bought Together