Order Entry
Czech Republic
ContactUsLinkComponent
 
Qualitative filter papers, grades MN 616 md and 616 md 1/4, circles and folded filters
 
undefined
Qualitative filter papers, grades MN 616 md and 616 md 1/4, circles and folded filters

 

  • Catalog No:
  • MANA533015
  • MANA533018
  • MANA533011
  • MANA533012
  • MANA533032
  • MANA533024
  • MANA533009
  • MANA433015
  • MANA433018
  • MANA533005
  • MANA433024
  • MANA533007
  • MANA433032
  • MANA433005
  • MANA433007
  • MANA433009
  • MANA433011
  • MANA433012
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • Particle retention:
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • 4 - 7 µm
  • Filtration speed:
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • 55 s (DIN 53137)
  • Ash content:
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%
  • ~0,1%

 

Frequently Bought Together