Anti-BCAT2 Rabbit Polyclonal Antibody
Catalog # ABNOH00000587-PW1
Supplier: Abnova
Specifications
- Antigen name:Branched Chain Amino-Acid Transaminase 2, Mitochondrial
- Antigen symbol:BCAT2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:587
- Antigen synonyms:BCAM|PP18|BCATM|BCT2
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-BCAT2 (300 ?l); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-BCAT2 (50 ?l)
Type:
Antigen: BCAT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human