Order Entry
Czech Republic
ContactUsLinkComponent
Tongs, ETFE
Tongs, ETFE
Catalog # ENTEC40-0315
Supplier:  ENTEGRIS
Tongs, ETFE
Catalog # ENTEC40-0315
Supplier:  ENTEGRIS

Specifications

  • Length:
    231 mm
  • Pk:
    4
  • Spanning area:
    152 - 254 mm

Specifications

About this item

ETFE, white.

  • For substrate handling
  • Hooked inward with squared end
  • Adjustable (4 positions)