Order Entry
Czech Republic
ContactUsLinkComponent
Tubings, PFA
Tubings, PFA
Catalog # 222-0000
Supplier:  ENTEGRIS
Tubings, PFA
Catalog # 222-0000
Supplier:  ENTEGRIS

Specifications

  • Tubing ext.Ø:
    3,180 mm
  • Tubing int.Ø:
    1,656 mm
  • Wall thickness:
    0,762 mm
  • Pk:
    1 Feet

Specifications

About this item

Made of PFA, 4100