Safety storage cabinets for chemicals, CS-Classic
: asecos
Storage of non-flammable chemicals and toxic substances in workroom.
- Robust construction and longevity – body made of sheet steel with highgrade structured paint finish
- No unauthorised use – doors lockable with cylinder locking (integration in a locking system possible)
- Easy transport – integrated transport base with removable, optional base cover
- Easy alignment – adjusting aids to compensate for uneven floor
- Five more door colours available upon request at no extra cost
: TÜV approved.
: Interior equipment packages need to be ordered separately (not included). Please check the website or contact Avantor for further information.
Frequently Bought Together