Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Nuclear Pore Complex Mouse Monoclonal Antibody [clone: 39C7]
Anti-Nuclear Pore Complex Mouse Monoclonal Antibody [clone: 39C7]
Catalog # BSENM-1397-250
Supplier:  Biosensis
undefined
Anti-Nuclear Pore Complex Mouse Monoclonal Antibody [clone: 39C7]
Catalog # BSENM-1397-250
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear Pore Complex
  • Clonality:
    Monoclonal
  • Clone:
    39C7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Yeast,
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    Nuclear pore complex protein Nup107
  • Immunogen:
    Yeast nuclear preparation
  • Size:
    250 µl
  • Pk:
    250 µl

Specifications

About this item

The Nuclear Core Complex (NPC) acts as a gateway for macromolecular traffic between the cytoplasm and the nucleus.

The specificity of this antibody has been confirmed by IC.
Application Information:
Immunocytochemistry (IC). A dilution of 1:50-1:500 is recommended for IC. This antibody does not work well for Western Blotting. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Nuclear Pore Complex
Clonality: Monoclonal
Clone: 39C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse, Yeast