Order Entry
Czech Republic
ContactUsLinkComponent
Anti-non-erythrocyte alpha-spectrin Mouse Monoclonal Antibody [clone: 3D7]
Anti-non-erythrocyte alpha-spectrin Mouse Monoclonal Antibody [clone: 3D7]
Catalog # BSENM-1575-100
Supplier:  Biosensis
undefined
Anti-non-erythrocyte alpha-spectrin Mouse Monoclonal Antibody [clone: 3D7]
Catalog # BSENM-1575-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    non-erythrocyte alpha-spectrin
  • Clonality:
    Monoclonal
  • Clone:
    3D7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    SPTAN1
  • Immunogen:
    This antibody was raised against a recombinant construct containing the seventh, eight and ninth repeats (amino acids 676-1043) of human alpha-II Spectrin. The 9th spectrin repeat also includes a Src-homology 3 domain. This construct was expressed in and purified from E. coli.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Spectrins are a family of filamentous cytoskeletal proteins that function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. The Spectrins form into dimers and further into tetramers of alpha and beta subunits (Ref: Entrez Gene). The alpha-II subunit is widely expressed in tissues but, in the nervous system, is found predominantly in neurons.

Antibody recognizes spectrin in multiple species. Approved applications are: WB, IHC(IF). Fixation is 4% PFA or frozen tissues, not yet tested in paraffin embedded tissues. Recommended dilution of 1:500-1,000 for IHC/IF and 1:5,000-10,000 for WB. The protein is seen as a major band at 240kDa depending on the species.
Application Information:
WB, IHC(IF). Recommended dilution of 1:500-1,000 for IHC/IF and 1:5,000-10,000 for WB. The protein is seen as a major band at 240kDa depending on the species.

Type: Primary
Antigen: non-erythrocyte alpha-spectrin
Clonality: Monoclonal
Clone: 3D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovineine, Pig, Rat