Specifications
- Antibody type:Primary
- Antigen name:Beta Lipotropin
- Antigen symbol:Beta-LPH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:Multiple epitopes because of the nature of the immunogen
- Cross adsorption:No
- Format:Lyophilized
- Immunogen:Synthetic peptide (179-267 aa) of human beta-Lipotropin conjugated to thyroglobulin was used as the immunogen.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Human beta-Lipotropin is a 93 amino acid polypeptide that is cleaved from carboxy-terminal fragment of the precursor pro-opiomelanocortin (POMC). It stimulates melanocytes to produce melanin, and can also be cleaved into smaller peptides including opioid peptides: gamma-lipotropin, alpha-MSH, beta-MSH, gamma-MSH, alpha-endorphin, beta-endorphin, gamma-endorphin and met-enkephalin
Antibody recognizes beta Lipotropin by native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Beta Lipotropin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat