ContactUsLinkComponent
Extractor for the perforator method, DIN EN 120
Extractor for the perforator method, DIN EN 120
Catalog # NEUB1050-11-1000
Extractor for the perforator method, DIN EN 120
Catalog # NEUB1050-11-1000
Specifications
  • Type:
    NS 71 flange
  • Flask capacity:
    1000 ml
  • Pk:
    1
Specifications
About this item
Complete assembly includes NS 71 flange, frit, stopcock, cooler, piston, erlenmeyer flask and ball pipe.