Order Entry
Czech Republic
ContactUsLinkComponent
Bouffant caps with visor, Hopen®
Bouffant caps with visor, Hopen®
Catalog # HOPE70.201S
Supplier:  Medicom
undefined
Bouffant caps with visor, Hopen®
Catalog # HOPE70.201S
Supplier:  Medicom

Specifications

  • Ø:
    530 mm
  • Garment application:
    Food
  • Colour:
    White
  • Description:
    Bouffant caps with visor, in polybag
  • Pk:
    100

Specifications

About this item

These disposable bouffant caps with visor made of PP are ideal for use in hygiene and food industries.

  • Cover the hair as required by HACCP standards
  • Elasticated for perfect fit
  • Lightweight fabric

Weight: 15 g/m²