pH meter, handheld, SensoDirect pH 110
LOVI721300
: Lovibond
- Description:pH meter, SensoDirect pH 110
- Seq:01
- Model:SensoDirect pH 110
- Connections:BNC
- Display:LCD
- Power supply:1 × 9 V battery
- Weight:0,380 kg
- W×D×H:110×34×208 mm
- pH accuracy:±0,07 (5 - 9 pH)
±0,1 (4 - 10 pH)
±0,2 (1 - 13 pH)
±0,2 (10,1 - 13 pH) - pH calibration:2-point
- pH range:0 - 14
- pH resolution:0,01
- Pk:1
The SensoDirect pH 110 is a simple, easy-to-use, portable pH meter.
- Manual calibration, large display and result hold function make analysis and result interpretation easy
- Equipped as standard with a rugged protective casing for field use
- With built-in electrode holder for safe storage
- Flip out stand for bench top use
- Supplied gel electrode is temperature resistant over the range 0 to 80 °C
- Fitted with a BNC connector as standard
Typical applications include: Drinking water, cooling/boiler water, waste water, pool water, surface water, water treatment companies, laboratories.
: Supplied with sturdy plastic case, battery, pH buffer (4,00/7,00), pH plastic electrode-type 110, guarantee sheet and instruction manual.