Order Entry
Czech Republic
ContactUsLinkComponent
Beard cover, Hopen®
Beard cover, Hopen®
Catalog # 113-8337
Supplier:  Medicom
undefined
Beard cover, Hopen®
Catalog # 113-8337
Supplier:  Medicom

Specifications

  • Packed:
    100/poly bag
  • Colour:
    White
  • Pk:
    100
  • Weight:
    1,5 g

Specifications

About this item

This disposable beard cover made of white PP covers the lower part of the face. It is HACCP approved and therefore ideal for use in food applications, such as catering and food processing. Also suitable for chemical, pharmaceutical and maintenance applications.

  • Sewn, elasticated
  • Latex-free

Weight: 1,5 g
Size: 45 cm (long, flat)