Anti-CACNA1G Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S178A-9]
Catalog # ABNOMAB11561
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, T type, alpha 1G subunit
- Antigen symbol:CACNA1G
- Clonality:Monoclonal
- Clone:S178A-9
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Liquid
- Gene ID:12291
- Antigen synonyms:Cav3.1d|[a]1G|mKIAA1123|a1G|alpha-1G
- Amino acid number:2052 to 2172
- Storage buffer:PBS, pH 7,4 (50% glycerol)
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant fusion protien corresponding to amino acids 2052-2172 at C-terminus of mouse Cacna1g.
- Purification:Protein G Purified
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant of Cacna1g.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: