Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC17A6 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S29-29]
Anti-SLC17A6 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S29-29]
Catalog # ABNOMAB11543
Supplier:  Abnova
Anti-SLC17A6 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S29-29]
Catalog # ABNOMAB11543
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute carrier family 17, member A6
  • Antigen symbol:
    SLC17A6
  • Clonality:
    Monoclonal
  • Clone:
    S29-29
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    84487
  • Antigen synonyms:
    Vglut2|Dnpi
  • Amino acid number:
    501 to 582
  • Storage buffer:
    PBS, pH 7,4 (50% glycerol)
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant fusion protien corresponding to amino acids 501-582 at C-terminus of rat Slc17a6.
  • Purification:
    Protein G Purified
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against recombinant of Slc17a6.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC17A6
Clonality: Monoclonal
Clone: S29-29
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: