Anti-FFAR2 Mouse Monoclonal Antibody [clone: 3B3]
Katalogové číslo ABNOH00002867-M02
Dodavatel: Abnova
Specifikace
- Antibody type:Primary
- Antigen name:Free fatty acid receptor 2
- Antigen symbol:FFAR2
- Clonality:Monoclonal
- Clone:3B3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2867
- Antigen synonyms:FFA2R|GPR43
- Amino acid number:231 to 330
- Storage buffer:1x PBS, pH 7,4
- Sequence:FLVCFGPYNVSHLVGYHQRKSPWWRSIAVVFSSLNASLDPLLFYFSSSVVRRAFGRGLQVLRNQGSSLLGRRGKDTAEGTNEDRGVGQGEGMPSSDFTTE
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:FFAR2 (NP_005297.1, 231 a.a. ~ 330 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 μg
- Pk:100 µg
Specifikace
O tomto produktu
Mouse monoclonal antibody raised against a partial recombinant FFAR2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: FFAR2
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: