Anti-HSPA1A Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 2A4]
Catalog # ABNOMAB11758
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 70kDA protein 1A
- Antigen symbol:HSPA1A
- Clonality:Monoclonal
- Clone:2A4
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3303
- Antigen synonyms:HSPA1|HSP70-1A|HSP72|HSP70-1|HSP70.1|HEL-S-103|HSP70I|Epididymis Secretory Protein Li 103
- Storage buffer:PBS, pH 7,2 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human HSPA1A.
- Purification:PEG Purification
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant HSPA1A.
Recommended Dilutions: Western Blot: 1:5000; Immunocytochemistry: 1:100; Immunofluorescence: 1:100; Immunohistochemistry: 1:100
Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 2A4
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: