Anti-CLCC1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25130
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chloride Channel CLIC-like 1
- Antigen symbol:CLCC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:23155
- Antigen synonyms:MCLC
- Storage buffer:PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to CLCC1.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of CLCC1.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunofluorescence: 1:50-1:200
Type: Primary
Antigen: CLCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human