Anti-CK8 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: CK8+18 207]
Catalog # ABNOMAB11168
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 8
- Antigen symbol:CK8
- Clonality:Monoclonal
- Clone:CK8+18 207
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:3856|3875
- Antigen synonyms:cytokeratin 8|Cytokeratin endo A|CYK8|Keratin|KRT8|type II cytoskeletal 8|K8|KRT2-8|CK 8|K2C8|CK8
- Storage buffer:PBS, pH 7,4 (1% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:Purified human KRT8/KRT18.
- Size:1 mL
- Pk:1 ml
Specifications
About this item
Mouse monoclonal antibody raised against KRT8/KRT18.
Recommended Dilutions: Immunocytochemistry: 1:50-1:100; Immunohistochemistry: 1:50-1:100; Immunofluorescence: Circ?lating Tumor Cell) (1:20
Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: CK8+18 207
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human