Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Hepatitis C virus NS5a (genotype 2A) Rabbit Polyclonal Antibody
Anti-Hepatitis C virus NS5a (genotype 2A) Rabbit Polyclonal Antibody
Catalog # ABNOPAB24633
Supplier:  Abnova
Anti-Hepatitis C virus NS5a (genotype 2A) Rabbit Polyclonal Antibody
Catalog # ABNOPAB24633
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Hepatitis C virus NS5a (genotype 2A)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    1x PBS, pH 7,2 (0,01% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to Hepatitis C virus NS5a, genotype 2A.
  • Purification:
    Peptide affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of Hepatitis C virus NS5a (genotype 2A).

Recommended Dilutions: ELISA: 1:2000; Western Blot: 1:2000

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: