Anti-U2AF2 Mouse Monoclonal Antibody [clone: 5G8]
ABNOH00011338-M03
: Abnova
- Antibody type:Primary
- Antigen name:U2 small nuclear RNA auxiliary factor 2
- Antigen symbol:U2AF2
- Clonality:Monoclonal
- Clone:5G8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:11338
- Antigen synonyms:U2AF65
- Amino acid number:141 to 240
- Storage buffer:1x PBS, pH 7,4
- Sequence:GSQMTRQARRLYVGNIPFGITEEAMMDFFNAQMRLGGLTQAPGNPVLAVQINQDKNFAFLEFRSVDETTQAMAFDGIIFQGQSLKIRRPHDYQPLPGMSE
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:U2AF2 (NP_001012496.1, 141 a.a. ~ 240 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against a partial recombinant U2AF2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: U2AF2
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human