Order Entry
Czech Republic
ContactUsLinkComponent
Anti-LATS2 Mouse Monoclonal Antibody [clone: ST-3B11]
Anti-LATS2 Mouse Monoclonal Antibody [clone: ST-3B11]
Catalog # ABNOMAB0009
Supplier:  Abnova
Anti-LATS2 Mouse Monoclonal Antibody [clone: ST-3B11]
Catalog # ABNOMAB0009
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Large tumor suppressor kinase 2
  • Antigen symbol:
    LATS2
  • Clonality:
    Monoclonal
  • Clone:
    ST-3B11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    26524
  • Antigen synonyms:
    KPM|Large tumor suppressor homolog 2|Serine/threonine-protein kinase kpm|LATS2|Kinase phosphorylated during mitosis protein|Serine/threonine-protein kinase LATS2|Warts-like kinase
  • Modification:
    phospho-Ser83
  • Storage buffer:
    10 mM HEPES/KOH, 150 mM NaCl, pH 7,5 (50% glycerol)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S83 of human LATS2.
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against synthetic phosphopeptide of LATS2.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; ELISA: 1 ?g/ml

Type: Primary
Antigen: LATS2
Clonality: Monoclonal
Clone: ST-3B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human