Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD9 Mouse Monoclonal Antibody [clone: MEM-61]
Anti-CD9 Mouse Monoclonal Antibody [clone: MEM-61]
Catalog # ABNOMAB0874
Supplier:  Abnova
Anti-CD9 Mouse Monoclonal Antibody [clone: MEM-61]
Catalog # ABNOMAB0874
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD9 molecule
  • Antigen symbol:
    CD9
  • Clonality:
    Monoclonal
  • Clone:
    MEM-61
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    928
  • Antigen synonyms:
    MIC3|TSPAN29|TSPAN-29|DRAP-27|MRP-1|BTCC-1
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified CD9 from human pre-B cell line NALM-6.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native CD9.

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: MEM-61
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human