Anti-TFRC Mouse Monoclonal Antibody [clone: MEM-75]
Catalog # ABNOMAB0897
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transferrin Receptor
- Antigen symbol:TFRC
- Clonality:Monoclonal
- Clone:MEM-75
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:7037
- Antigen synonyms:TRFR|T9|TFR1|CD71|p90|TFR|TR
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified TFRC from human pre-B cell line NALM-6.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native TFRC.
Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: MEM-75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human