Anti-CASP8 Mouse Monoclonal Antibody [clone: FLICE 1 1-37]
ABNOMAB1844
: Abnova
- Antibody type:Primary
- Antigen name:Caspase 8
- Antigen symbol:CASP8
- Clonality:Monoclonal
- Clone:FLICE 1 1-37
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:841
- Antigen synonyms:FLICE|caspase 8|MACH-beta-1/2/3/4 protein|FADD-homologous ICE/CED-3-like protease|MCH5|apoptotic protease Mch-5|FADD-like ICE|MACH-alpha-1/2/3 protein|CAP4|ALPS2B|apoptotic cysteine protease|Casp-8|MORT1-associated ced-3 homolog|caspase-8|apoptosis-related cysteine protease|ICE-like apoptotic protease 5|MACH
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CASP8.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against recombinant CASP8.
Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: FLICE 1 1-37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human