Anti-CYP2A6 Mouse Monoclonal Antibody [clone: F16 P2 D8]
Catalog # ABNOMAB1858
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 2, Subfamily A, Polypeptide 6
- Antigen symbol:CYP2A6
- Clonality:Monoclonal
- Clone:F16 P2 D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1548
- Antigen synonyms:P450C2A|CPA6|CYPIIA6|P450PB|CYP2A3|CYP2A
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of human CYP2A6.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of CYP2A6.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: CYP2A6
Clonality: Monoclonal
Clone: F16 P2 D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human